Skip to Content
Merck
Tryptophan European Pharmacopoeia (EP) Reference Standard

Tryptophan

Tryptophan European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(S)-2-Amino-3-(3-indolyl)propionic acid, L-α-Amino-3-indolepropionic acid, L-Tryptophan
Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
API Family
Reference Material Type
Special Grade
Technique