Merck
GC Analysis of Noble Gases on Carboxen®-1010 PLOT
GC Analysis of Noble Gases on Carboxen®-1010 PLOT suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Carboxen®-1010 PLOT Capillary GC Column
L × I.D. 30 m × 0.53 mm, average thickness 30 μm
CONDITIONS
column
Carboxen-1010 PLOT, 30 m × 0.53 mm I.D. (25467)
oven
35 °C (6 min), 24 °C/min to 225 °C
detector
TCD
carrier gas
helium, 3 mL/min. @ 35 °C
Description
Legal Information
Carboxen is a registered trademark of Merck KGaA, Darmstadt, Germany