Merck
  • A patient with germ-line gain-of-function PDGFRB p.N666H mutation and marked clinical response to imatinib.
A patient with germ-line gain-of-function PDGFRB p.N666H mutation and marked clinical response to imatinib.
Genetics in medicine : official journal of the American College of Medical Genetics (2017-07-21)
Dinel Pond, Florence A Arts, Nancy J Mendelsohn, Jean-Baptiste Demoulin, Gunter Scharer, Yoav Messinger
ABSTRACT
PurposeHeterozygous germ-line activating mutations in PDGFRB cause Kosaki and Penttinen syndromes and myofibromatosis. We describe a 10-year-old child with a germ-line PDGFRB p.N666H mutation who responded to the tyrosine kinase inhibitor imatinib by inhibition of PDGFRB.MethodsThe impact of p.N666H on PDGFRB function and sensitivity to imatinib was studied in cell culture.ResultsCells expressing the p.N666H mutation showed constitutive PDGFRB tyrosine phosphorylation. PDGF-independent proliferation was abolished by imatinib at 1 μM concentration. Patient fibroblasts showed constitutive receptor tyrosine phosphorylation that was also abrogated by imatinib with reduced proliferation of treated cells.This led to patient treatment with imatinib at 400 mg daily (340 mg/m