Discovery and refinement of loci associated with lipid levels.
Nature genetics (2013-10-08)
Cristen J Willer, Ellen M Schmidt, Sebanti Sengupta, Gina M Peloso, Stefan Gustafsson, Stavroula Kanoni, Andrea Ganna, Jin Chen, Martin L Buchkovich, Samia Mora, Jacques S Beckmann, Jennifer L Bragg-Gresham, Hsing-Yi Chang, Ayşe Demirkan, Heleen M Den Hertog, Ron Do, Louise A Donnelly, Georg B Ehret, Tõnu Esko, Mary F Feitosa, Teresa Ferreira, Krista Fischer, Pierre Fontanillas, Ross M Fraser, Daniel F Freitag, Deepti Gurdasani, Kauko Heikkilä, Elina Hyppönen, Aaron Isaacs, Anne U Jackson, Åsa Johansson, Toby Johnson, Marika Kaakinen, Johannes Kettunen, Marcus E Kleber, Xiaohui Li, Jian'an Luan, Leo-Pekka Lyytikäinen, Patrik K E Magnusson, Massimo Mangino, Evelin Mihailov, May E Montasser, Martina Müller-Nurasyid, Ilja M Nolte, Jeffrey R O'Connell, Cameron D Palmer, Markus Perola, Ann-Kristin Petersen, Serena Sanna, Richa Saxena, Susan K Service, Sonia Shah, Dmitry Shungin, Carlo Sidore, Ci Song, Rona J Strawbridge, Ida Surakka, Toshiko Tanaka, Tanya M Teslovich, Gudmar Thorleifsson, Evita G Van den Herik, Benjamin F Voight, Kelly A Volcik, Lindsay L Waite, Andrew Wong, Ying Wu, Weihua Zhang, Devin Absher, Gershim Asiki, Inês Barroso, Latonya F Been, Jennifer L Bolton, Lori L Bonnycastle, Paolo Brambilla, Mary S Burnett, Giancarlo Cesana, Maria Dimitriou, Alex S F Doney, Angela Döring, Paul Elliott, Stephen E Epstein, Gudmundur Ingi Eyjolfsson, Bruna Gigante, Mark O Goodarzi, Harald Grallert, Martha L Gravito, Christopher J Groves, Göran Hallmans, Anna-Liisa Hartikainen, Caroline Hayward, Dena Hernandez, Andrew A Hicks, Hilma Holm, Yi-Jen Hung, Thomas Illig, Michelle R Jones, Pontiano Kaleebu, John J P Kastelein, Kay-Tee Khaw, Eric Kim, Norman Klopp, Pirjo Komulainen, Meena Kumari, Claudia Langenberg, Terho Lehtimäki, Shih-Yi Lin, Jaana Lindström, Ruth J F Loos, François Mach, Wendy L McArdle, Christa Meisinger, Braxton D Mitchell, Gabrielle Müller, Ramaiah Nagaraja, Narisu Narisu, Tuomo V M Nieminen, Rebecca N Nsubuga, Isleifur Olafsson, Ken K Ong, Aarno Palotie, Theodore Papamarkou, Cristina Pomilla, Anneli Pouta, Daniel J Rader, Muredach P Reilly, Paul M Ridker, Fernando Rivadeneira, Igor Rudan, Aimo Ruokonen, Nilesh Samani, Hubert Scharnagl, Janet Seeley, Kaisa Silander, Alena Stančáková, Kathleen Stirrups, Amy J Swift, Laurence Tiret, Andre G Uitterlinden, L Joost van Pelt, Sailaja Vedantam, Nicholas Wainwright, Cisca Wijmenga, Sarah H Wild, Gonneke Willemsen, Tom Wilsgaard, James F Wilson, Elizabeth H Young, Jing Hua Zhao, Linda S Adair, Dominique Arveiler, Themistocles L Assimes, Stefania Bandinelli, Franklyn Bennett, Murielle Bochud, Bernhard O Boehm, Dorret I Boomsma, Ingrid B Borecki, Stefan R Bornstein, Pascal Bovet, Michel Burnier, Harry Campbell, Aravinda Chakravarti, John C Chambers, Yii-Der Ida Chen, Francis S Collins, Richard S Cooper, John Danesh, George Dedoussis, Ulf de Faire, Alan B Feranil, Jean Ferrières, Luigi Ferrucci, Nelson B Freimer, Christian Gieger, Leif C Groop, Vilmundur Gudnason, Ulf Gyllensten, Anders Hamsten, Tamara B Harris, Aroon Hingorani, Joel N Hirschhorn, Albert Hofman, G Kees Hovingh, Chao Agnes Hsiung, Steve E Humphries, Steven C Hunt, Kristian Hveem, Carlos Iribarren, Marjo-Riitta Järvelin, Antti Jula, Mika Kähönen, Jaakko Kaprio, Antero Kesäniemi, Mika Kivimaki, Jaspal S Kooner, Peter J Koudstaal, Ronald M Krauss, Diana Kuh, Johanna Kuusisto, Kirsten O Kyvik, Markku Laakso, Timo A Lakka, Lars Lind, Cecilia M Lindgren, Nicholas G Martin, Winfried März, Mark I McCarthy, Colin A McKenzie, Pierre Meneton, Andres Metspalu, Leena Moilanen, Andrew D Morris, Patricia B Munroe, Inger Njølstad, Nancy L Pedersen, Chris Power, Peter P Pramstaller, Jackie F Price, Bruce M Psaty, Thomas Quertermous, Rainer Rauramaa, Danish Saleheen, Veikko Salomaa, Dharambir K Sanghera, Jouko Saramies, Peter E H Schwarz, Wayne H-H Sheu, Alan R Shuldiner, Agneta Siegbahn, Tim D Spector, Kari Stefansson, David P Strachan, Bamidele O Tayo, Elena Tremoli, Jaakko Tuomilehto, Matti Uusitupa, Cornelia M van Duijn, Peter Vollenweider, Lars Wallentin, Nicholas J Wareham, John B Whitfield, Bruce H R Wolffenbuttel, Jose M Ordovas, Eric Boerwinkle, Colin N A Palmer, Unnur Thorsteinsdottir, Daniel I Chasman, Jerome I Rotter, Paul W Franks, Samuli Ripatti, L Adrienne Cupples, Manjinder S Sandhu, Stephen S Rich, Michael Boehnke, Panos Deloukas, Sekar Kathiresan, Karen L Mohlke, Erik Ingelsson, Gonçalo R Abecasis