Merck
  • Deleterious mutation in FDX1L gene is associated with a novel mitochondrial muscle myopathy.
Deleterious mutation in FDX1L gene is associated with a novel mitochondrial muscle myopathy.
European journal of human genetics : EJHG (2013-11-28)
Ronen Spiegel, Ann Saada, Jonatan Halvardson, Devorah Soiferman, Avraham Shaag, Simon Edvardson, Yoseph Horovitz, Morad Khayat, Stavit A Shalev, Lars Feuk, Orly Elpeleg
ABSTRACT
Isolated metabolic myopathies encompass a heterogeneous group of disorders, with mitochondrial myopathies being a subgroup, with depleted skeletal muscle energy production manifesting either by recurrent episodes of myoglobinuria or progressive muscle weakness. In this study, we investigated the genetic cause of a patient from a consanguineous family who presented with adolescent onset autosomal recessive mitochondrial myopathy. Analysis of enzyme activities of the five respiratory chain complexes in our patients' skeletal muscle showed severely impaired activities of iron sulfur (Fe-S)-dependent complexes I, II and III and mitochondrial aconitase. We employed exome sequencing combined with homozygosity mapping to identify a homozygous mutation, c.1A>T, in the FDX1L gene, which encodes the mitochondrial ferredoxin 2 (Fdx2) protein. The mutation disrupts the ATG initiation translation site resulting in severe reduction of Fdx2 content in the patient muscle and fibroblasts mitochondria. Fdx2 is the second component of the Fe-S cluster biogenesis machinery, the first being IscU that is associated with isolated mitochondrial myopathy. We suggest adding genetic analysis of FDX1L in cases of mitochondrial myopathy especially when associated with reduced activity of the respiratory chain complexes I, II and III.