Merck
  • Actin-binding protein 1 links B-cell antigen receptors to negative signaling pathways.
Actin-binding protein 1 links B-cell antigen receptors to negative signaling pathways.
Proceedings of the National Academy of Sciences of the United States of America (2014-06-25)
Margaret K Seeley-Fallen, Lisa J Liu, Melanie R Shapiro, Olusegun O Onabajo, Senthilkumar Palaniyandi, Xiaoping Zhu, Tse-Hua Tan, Arpita Upadhyaya, Wenxia Song
ABSTRACT
Prolonged or uncontrolled B-cell receptor (BCR) signaling is associated with autoimmunity. We previously demonstrated a role for actin in BCR signal attenuation. This study reveals that actin-binding protein 1 (Abp1/HIP-55/SH3P7) is a negative regulator of BCR signaling and links actin to negative regulatory pathways of the BCR. In both Abp1(-/-) and bone marrow chimeric mice, in which only B cells lack Abp1 expression, the number of spontaneous germinal center and marginal zone B cells and the level of autoantibody are significantly increased. Serum levels of T-independent antibody responses and total antibody are elevated, whereas T-dependent antibody responses are markedly reduced and fail to undergo affinity maturation. Upon activation, surface BCR clustering is enhanced and B-cell contraction delayed in Abp1(-/-) B cells, concurrent with slow but persistent increases in F-actin at BCR signalosomes. Furthermore, BCR signaling is enhanced in Abp1(-/-) B cells compared with wild-type B cells, including Ca(2+) flux and phosphorylation of B-cell linker protein, the mitogen-activated protein kinase kinase MEK1/2, and ERK, coinciding with reductions in recruitment of the inhibitory signaling molecules hematopoietic progenitor kinase 1 and SH2-containing inositol 5-phosphatase to BCR signalosomes. Our results indicate that Abp1 negatively regulates BCR signaling by coupling actin remodeling to B-cell contraction and activation of inhibitory signaling molecules, which contributes to the regulation of peripheral B-cell development and antibody responses.
Sigma-Aldrich
Fructose-6-phosphate Kinase from Bacillus stearothermophilus, Type VII, lyophilized powder, ≥50 units/mg protein