Merck
  • Tranilast prevents atrial remodeling and development of atrial fibrillation in a canine model of atrial tachycardia and left ventricular dysfunction.
Tranilast prevents atrial remodeling and development of atrial fibrillation in a canine model of atrial tachycardia and left ventricular dysfunction.
Journal of the American College of Cardiology (2013-01-01)
Yosuke Nakatani, Kunihiro Nishida, Masao Sakabe, Naoya Kataoka, Tamotsu Sakamoto, Yoshiaki Yamaguchi, Jotaro Iwamoto, Koichi Mizumaki, Akira Fujiki, Hiroshi Inoue
ABSTRACT
This study sought to assess the effects of tranilast on atrial remodeling in a canine atrial fibrillation (AF) model. Tranilast inhibits transforming growth factor (TGF)-β1 and prevents fibrosis in many pathophysiological settings. However, the effects of tranilast on atrial remodeling remain unclear. Beagles were subjected to atrial tachypacing (400 beats/min) for 4 weeks while treated with placebo (control dogs, n = 8) or tranilast (tranilast dogs, n = 10). Sham dogs (n = 6) did not receive atrial tachypacing. Atrioventricular conduction was preserved. Ventricular dysfunction developed in the control and tranilast dogs due to rapid ventricular responses. Atrial fibrillation duration (211 ± 57 s) increased, and AF cycle length and atrial effective refractory period shortened in controls, but these changes were suppressed in tranilast dogs (AF duration, 18 ± 10 s, p < 0.01 vs. control). The L-type calcium channel α1c (Cav1.2) micro ribonucleic acid expression decreased in control dogs (sham 1.38 ± 0.24 vs. control 0.65 ± 0.12, p < 0.01), but not in tranilast dogs (0.97 ± 0.14, p = not significant vs. sham). Prominent atrial fibrosis (fibrous tissue area, sham 0.8 ± 0.1 vs. control 9.3 ± 1.3%, p < 0.01) and increased expression of tissue inhibitor of metalloproteinase protein 1 were observed in control dogs but not in tranilast dogs (fibrous tissue area, 1.4 ± 0.2%, p < 0.01 vs. control). The TGF-β1 (sham 1.00 ± 0.07 vs. control 3.06 ± 0.87, p < 0.05) and Rac1 proteins were overexpressed in control dogs, but their overexpression was inhibited in tranilast dogs (TGF-β1, 1.28 ± 0.20, p < 0.05 vs. control). Tranilast prevented atrial remodeling and suppressed AF development in a canine model. Its inhibition of TGF-β1 and Rac1 overexpression may contribute to its antiremodeling effects.
Sigma-Aldrich
Tranilast, ≥98% (HPLC), powder