Merck
  • Syntheses of carbocyclic analogues of α-D-glucosamine, α-D-mannose, α-D-mannuronic acid, β-L-idosamine, and β-L-gulose.
Syntheses of carbocyclic analogues of α-D-glucosamine, α-D-mannose, α-D-mannuronic acid, β-L-idosamine, and β-L-gulose.
The Journal of organic chemistry (2012-08-03)
Yunshan Sun, Mark Nitz
ABSTRACT
A versatile synthesis of orthogonally protected derivatives of carba-α-D-glucosamine, carba-α-D-mannose, carba-α-D-mannuronic acid, carba-β-L-idosamine, and carba-β-L-gulose from methyl α-D-mannoside is described. Our synthetic strategy utilizes the palladium-promoted Ferrier carbocyclization and persistent butane-2,3-diacetal protection to produce a key chiral cyclohexanone intermediate, from which all five carbasugar derivatives can readily be obtained.