Merck
  • Combined fluorescent-chromogenic in situ hybridization for identification and laser microdissection of interphase chromosomes.
Combined fluorescent-chromogenic in situ hybridization for identification and laser microdissection of interphase chromosomes.
PloS one (2013-04-09)
Nerea Paz, Amaia Zabala, Félix Royo, África García-Orad, José L Zugaza, Luis A Parada
ABSTRACT
Chromosome territories constitute the most conspicuous feature of nuclear architecture, and they exhibit non-random distribution patterns in the interphase nucleus. We observed that in cell nuclei from humans with Down Syndrome two chromosomes 21 frequently localize proximal to one another and distant from the third chromosome. To systematically investigate whether the proximally positioned chromosomes were always the same in all cells, we developed an approach consisting of sequential FISH and CISH combined with laser-microdissection of chromosomes from the interphase nucleus and followed by subsequent chromosome identification by microsatellite allele genotyping. This approach identified proximally positioned chromosomes from cultured cells, and the analysis showed that the identity of the chromosomes proximally positioned varies. However, the data suggest that there may be a tendency of the same chromosomes to be positioned close to each other in the interphase nucleus of trisomic cells. The protocol described here represents a powerful new method for genome analysis.
Sigma-Aldrich
GenomePlex® Single Cell Whole Genome Amplification Kit, Amplify genome of a single cell
Sigma-Aldrich
GenomePlex® WGA Reamplification Kit, Reamplification of WGA product with minimal bias