Merck
  • Insulinoma-associated protein 1 controls nasopharyngeal carcinoma to radiotherapy by modulating cyclin D1-dependent DNA repair machinery.
Insulinoma-associated protein 1 controls nasopharyngeal carcinoma to radiotherapy by modulating cyclin D1-dependent DNA repair machinery.
Carcinogenesis (2019-06-04)
Ruibiao Song, Xing Wei, You Wang, Shousen Hu, Yunpeng Ba, Xiyan Xiao, Jianzhong Zhang
ABSTRACT
Insulinoma-associated protein 1 (INSM1), a zinc finger transcriptional factor, is proven to be deregulated in several types of cancers. However, comprehension of the molecular mechanism of INSM1-mediated tumor progression remains poor. Here, we show that the radioresistant nasopharyngeal carcinoma (NPC) patients have higher expressions of INSM1 that correlated with poor prognosis. Genetic manipulation of INSM1 expression sufficiently controls the response of NPC cells to irradiation (IR). Mechanistically, cells exposed to IR, increased intracellular INSM1 competitively disrupts the interaction of cyclin D1 and CDK4 resulting in cell survival by the cyclin D1-dependent DNA repair machinery. Moreover, knockdown of INSM1 sensitives NPC cells to IR in vivo and protects xenograft mice from mortality. Taken together, these results indicate that INSM1 modulates NPC to radiotherapy by controlling cyclin D1-dependent DNA repair machinery that could be manipulated as a novel molecular target for NPC therapy.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin fragment 26-33 Amide, ≥97% (HPLC), powder