Merck
  • Reliability of Docking-Based Virtual Screening for GPCR Ligands with Homology Modeled Structures: A Case Study of the Angiotensin II Type I Receptor.
Reliability of Docking-Based Virtual Screening for GPCR Ligands with Homology Modeled Structures: A Case Study of the Angiotensin II Type I Receptor.
ACS chemical neuroscience (2018-09-29)
Haiyi Chen, Weitao Fu, Zhe Wang, Xuwen Wang, Tailong Lei, Feng Zhu, Dan Li, Shan Chang, Lei Xu, Tingjun Hou
ABSTRACT
The number of solved G-protein-coupled receptor (GPCR) crystal structures has expanded rapidly, but most GPCR structures remain unsolved. Therefore, computational techniques, such as homology modeling, have been widely used to produce the theoretical structures of various GPCRs for structure-based drug design (SBDD). Due to the low sequence similarity shared by the transmembrane domains of GPCRs, accurate prediction of GPCR structures by homology modeling is quite challenging. In this study, angiotensin II type I receptor (AT1R) was taken as a typical case to assess the reliability of class A GPCR homology models for SBDD. Four homology models of angiotensin II type I receptor (AT1R) at the inactive state were built based on the crystal structures of CXCR4 chemokine receptor, CCR5 chemokine receptor, and δ-opioid receptor, and refined through molecular dynamics (MD) simulations and induced-fit docking, to allow for backbone and side-chain flexibility. Then, the quality of the homology models was assessed relative to the crystal structures in terms of two criteria commonly used in SBDD: prediction accuracy of ligand-binding poses and screening power of docking-based virtual screening. It was found that the crystal structures outperformed the homology models prior to any refinement in both assessments. MD simulations could generally improve the docking results for both the crystal structures and homology models. Moreover, the optimized homology model refined by MD simulations and induced-fit docking even shows a similar performance of the docking assessment to the crystal structures. Our results indicate that it is possible to establish a reliable class A GPCR homology model for SBDD through the refinement by integrating multiple molecular modeling techniques.