Merck
  • Ancestral T Cells in Fish Require mTORC1-Coupled Immune Signals and Metabolic Programming for Proper Activation and Function.
Ancestral T Cells in Fish Require mTORC1-Coupled Immune Signals and Metabolic Programming for Proper Activation and Function.
Journal of immunology (Baltimore, Md. : 1950) (2019-07-28)
Xiumei Wei, Kete Ai, Huiying Li, Yu Zhang, Kang Li, Jialong Yang
ABSTRACT
T cells suddenly appeared in jawed fish ∼450 million years ago. Biological studies of fish T cells may provide helpful evidence to understand evolution of adaptive immune systems. To this end, using a Nile tilapia (Oreochromis niloticus) model, we revealed the regulatory mechanism of adaptive immunity mediated by ancestral T cells in jawed fish. Nile tilapia T cells as well as a tightly regulated mammalian/mechanistic target of rapamycin complex 1 (mTORC1) pathway participate in the cellular adaptive immune response during Streptococcus agalactiae infection. Blockade of mTORC1 signaling by rapamycin impairs T cell activation and Ag-induced proliferation in this early vertebrate. More critically, we show that signals from mTORC1 are indispensable for primordial effector T cells to eliminate infection by promoting the expression of proinflammatory cytokines, cytotoxic-related molecules, and proapoptotic genes. Mechanistically, teleost mTORC1 directs effector T cell function by coordinating multiple metabolic programs, including glycolysis, glutaminolysis, and lipogenesis through activating key transcription factors c-Myc, HIF-1α, and sterol regulatory element-binding proteins, and thus links immune signals to metabolic reprogramming in jawed fish. To our knowledge, these results represent the first description of the regulatory mechanism for T cell-mediated adaptive immunity in a fish species. From an evolutionary viewpoint, our study suggests that primordial T cells are armed with sophisticated regulatory strategies like those in modern T cells prior to the divergence of bony fish from the tetrapod lineage. Therefore, our findings fill in an important gap regarding evolution of the adaptive immune system.