Skip to Content
NO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whatman
®
ME membranes for microbiological control
Products
Product Category
filter discs (26)
Manufacturer
Diameter (mm)
Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-20 of 26
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
WHA10406972
Whatman
®
ME membranes for microbiological control,
ME24/21 ST, diam. 50 mm, pore size 0.2 μm, white with black grid, sterile
Expand
View Pricing
WHA10406970
Whatman
®
ME membranes for microbiological control,
ME24/21, diam. 47 mm, pore size 0.2 μm, white with black grid
Expand
View Pricing
WHA10406871
Whatman
®
ME membranes for microbiological control,
ME25/21, diam. 47 mm, pore size 0.45 μm, white with black grid
Expand
View Pricing
WHA10407312
Whatman
®
ME membranes for microbiological control,
ME25/21 STL membrane circles white, diam. 47 mm, pore size 0.45 μm, black grid, sterile
Expand
View Pricing
WHA10407970
Whatman
®
ME membranes for microbiological control,
ME25/41 ST, diam. 47 mm, pore size 0.45 μm, white with black grid, sterile
Expand
View Pricing
WHA10409770
Whatman
®
ME membranes for microbiological control,
ME25/31 STL, diam. 47 mm, pore size 0.45 μm, black with white grid, sterile
Expand
View Pricing
WHA10408970
Whatman
®
ME membranes for microbiological control,
ME27/21 STL, 3.1 mm grid, for Membrane-Butler, pore size 0.8 μm, black grid, diam. 47 mm
Expand
View Pricing
WHA10409970
Whatman
®
ME membranes for microbiological control,
ME27/61, 3.1 mm grid, pore size 0.8 μm, black grid, diam. 47 mm
Expand
View Pricing
WHA10408714
Whatman
®
ME membranes for microbiological control,
ME24/21 ST membrane circle white, diam. 50 mm, pore size 0.2 μm, black grid, sterile
Expand
View Pricing
WHA10406512
Whatman
®
ME membranes for microbiological control,
ME25/20, diam. 47 mm, pore size 0.45 μm, white with black grid
Expand
View Pricing
WHA10408472
Whatman
®
ME membranes for microbiological control,
ME28/41 ST, diam. 50 mm, pore size 1.2 μm, black grid, sterile
Expand
View Pricing
WHA10409470
Whatman
®
ME membranes for microbiological control,
ME25/41 ST membrane circles green, diam. 47 mm, pore size 0.45 μm, black grid, sterile
Expand
View Pricing
WHA10407314
Whatman
®
ME membranes for microbiological control,
ME25/21 STL membrane circles white, diam. 50 mm, pore size 0.45 μm, black grid, sterile
Expand
View Pricing
WHA10406572
Whatman
®
ME membranes for microbiological control,
ME20/25, 5 mm grid, pore size 0.45 μm, black grid (5 mm grid), diam. 50 mm
Expand
View Pricing
WHA10407332
Whatman
®
ME membranes for microbiological control,
ME25/31 STL membrane circles black, diam. 47 mm, pore size 0.45 μm, white grid, sterile
Expand
View Pricing
WHA10408712
Whatman
®
ME membranes for microbiological control,
ME24/21, without interleaving paper, pore size 0.2 μm, black grid, diam. 50 mm
Expand
View Pricing
WHA10409834
Whatman
®
ME membranes for microbiological control,
ME26/31, 3.1 mm grid, for Membrane-Butler, pore size 0.6 μm, white grid, diam. 50 mm
Expand
View Pricing
WHA10406812
Whatman
®
ME membranes for microbiological control,
ME25/21, diam. 47 mm, pore size 0.45 μm, white with black grid
Expand
View Pricing
WHA10407372
Whatman
®
ME membranes for microbiological control,
ME25/41 STL membrane circles green, diam. 50 mm, pore size 0.45 μm, black grid, sterile
Expand
View Pricing
WHA10407334
Whatman
®
ME membranes for microbiological control,
ME25/31 STL membrane circle black, diam. 50 mm, pore size 0.45 μm, white grid, sterile
Expand
View Pricing
Page 1 of 2
Page 1 of 2