Skip to Content
Merck
Deferoxamine for system suitability European Pharmacopoeia (EP) Reference Standard

Deferoxamine mesylate

Deferoxamine for system suitability European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Deferoxamine mesylate salt, DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt
Empirical Formula (Hill Notation):
C25H48N6O8 · CH4O3S
CAS Number:
Molecular Weight:
656.79

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade