Skip to Content
Merck
Trolamine impurity A European Pharmacopoeia (EP) Reference Standard

Monoethanolamine

Trolamine impurity A European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Ethanolamine, 2-Aminoethanol, 2-Aminoethyl alcohol, ETA, MEA, MEA 90, MEA-LCI, Monoethanolamine
Linear Formula:
NH2CH2CH2OH
CAS Number:
Molecular Weight:
61.08

Products

Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Technique