Skip to Content
Merck
N-Acetylglucosamine United States Pharmacopeia (USP) Reference Standard

N-Acetylglucosamine

N-Acetylglucosamine United States Pharmacopeia (USP) Reference Standard
Synonym(s):
N-Acetyl-D-glucosamine, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc
Empirical Formula (Hill Notation):
C8H15NO6
CAS Number:
Molecular Weight:
221.21

Products

Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique