Benzalkonium chloride solution
Synonym(s):
Alkylbenzyldimethylammonium chloride
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Product Line
Agency
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing