Merck
GC Analysis of Active Amyl Alcohol, Isoamyl Alcohol, and C10-C14 n-Alkane Markers on SLB®-IL76i
GC Analysis of Active Amyl Alcohol, Isoamyl Alcohol, and C10-C14 n-Alkane Markers on SLB®-IL76i suitable for GC
Materials
analytical column
Product No.
Description
Pricing
SLB®-IL76i Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.20 μm
CONDITIONS
column
SLB-IL76i, 30 m x 0.25 mm I.D., 0.20 μm (29869-U)
oven
90 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
hydrogen, 40 cm/sec
injection
1 μL, 100:1 split
liner
4 mm I.D., split type, cup design
sample
7 analytes in methylene chloride
Description
Analysis Note
Active amyl alcohol and isoamyl alcohol are by-products of the fermentation process. They are of concern to alcoholic beverage manufacturers due to their aroma characteristics. In this application, both analytes and several n-alkane markers are analyzed on SLB IL76i. This inert ionic liquic column produces great peak shapes for both alcohols, resulting in great resolution.
Legal Information
SLB is a registered trademark of Merck KGaA, Darmstadt, Germany