Merck
  • Photochemistry of N-methylformamide: matrix isolation and nonadiabatic dynamics.
Photochemistry of N-methylformamide: matrix isolation and nonadiabatic dynamics.
Chemphyschem : a European journal of chemical physics and physical chemistry (2012-10-04)
Rachel Crespo-Otero, Artur Mardyukov, Elsa Sanchez-Garcia, Mario Barbatti, Wolfram Sander
ABSTRACT
The photochemistry of N-methylformamide (MF) is elucidated by investigating its photodissociation products generated by UV irradiation (248 nm) in an argon matrix (10 K). We find that, starting from trans-MF, prolonged irradiation produces cis-MF, CH3NH2 and CO fragments as major products. Another photoproduct is identified as methylformimidic acid (FIA). Nonadiabatic dynamics simulations starting from both MF conformers revealed that the internal conversion occurs within 1 ps through a C-N dissociation channel. The major product is a weakly bound complex between CH3NH and HCO radicals. This complex owes its existence to the cage effect of the matrix which allows for H-transfer reactions and recombination. By identifying the primary photoisomerization and photodissociation pathways of MF, we gain new insights into the photochemistry of peptide bonds in general, which is a prerequisite for a better understanding of the effect of UV irradiation on living systems.
Sigma-Aldrich
N-Methylformamide, 99%
Sigma-Aldrich
N-Methylformamide, 99%