Merck
  • Study of dried blood spots technique for the determination of dextromethorphan and its metabolite dextrorphan in human whole blood by LC-MS/MS.
Study of dried blood spots technique for the determination of dextromethorphan and its metabolite dextrorphan in human whole blood by LC-MS/MS.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2009-02-25)
Xiaorong Liang, Yinghe Li, Matthew Barfield, Qin C Ji
ABSTRACT
Dried blood spots (DBSs) technology was evaluated in an assay for the quantitation of dextromethorphan (DM) and its metabolite, dextrorphan (DT), in human whole blood using high performance liquid chromatography with tandem mass spectrometry method (LC-MS/MS). Both the parent drug and metabolite were spiked in the blood matrix and subsequently allowed to dry on a specimen collection card. The dried blood spots were removed using a manual punch and then extracted into methyl tert-butyl ether (MTBE). The organic supernatant was transferred and evaporated and the residue was reconstituted in 20% acetonitrile. The overall method recovery of DM and DT was 87.8% and 95.4%, respectively. The assay was linear over the concentration range of 0.2-200ng/mL for both analytes. Several factors that potentially affect DBS assay quantitation were investigated, such as punch size, DBS sample punch-out location, and the volume of the blood sample pipetted on the specimen collection cards. The study determined that punch size does not affect assay quantitation accuracy. Indeed, a larger punch size increases the sensitivity due to the larger sampled blood spots. Sampling from different location on the specimen collection cards shows no significant variation for both drugs. The study also shows that acceptable results can be achieved with some variation of the sample volume, which allows a simple blood sampling procedure at the test sites. To achieve the similar lower limit of quantitation (LLOQ) as the plasma assay, several blood spots at the same concentration level were stacked together and extracted. Bioanalytical assays using the DBS technique are promising given the advantages of the method over the plasma assay.