Merck
  • Functional characterization of three naturally occurring single nucleotide polymorphisms in the CES2 gene encoding carboxylesterase 2 (HCE-2).
Functional characterization of three naturally occurring single nucleotide polymorphisms in the CES2 gene encoding carboxylesterase 2 (HCE-2).
Drug metabolism and disposition: the biological fate of chemicals (2005-07-22)
Takashi Kubo, Su-Ryang Kim, Kimie Sai, Yoshiro Saito, Toshiharu Nakajima, Kenji Matsumoto, Hirohisa Saito, Kuniaki Shirao, Noboru Yamamoto, Hironobu Minami, Atsushi Ohtsu, Teruhiko Yoshida, Nagahiro Saijo, Yasuo Ohno, Shogo Ozawa, Jun-Ichi Sawada
ABSTRACT
Twelve single nucleotide polymorphisms (SNPs) in the human CES2 gene, which encodes a carboxylesterase, hCE-2 [human carboxylesterase 2 (EC 3.1.1.1)], have been reported in the Japanese. In this report, we have examined functional alterations of three SNPs, a nonsynonymous SNP (100C>T, R34W), an SNP at the splice acceptor site in intron 8 (IVS8-2A>G), and one newly discovered nonsynonymous SNP (424G>A, V142M). For the two nonsynonymous SNPs, the corresponding variant cDNAs were expressed in COS-1 cells. Both the R34W and V142M variants showed little esterase activities toward the anticancer agent irinotecan and two typical carboxylesterase substrates, p-nitrophenol acetate and 4-methylumbelliferyl acetate, although increased levels of cDNA-mediated protein expression were observed by Western blotting as compared with the wild type. To investigate a possible splicing aberration in IVS8-2A>G, an in vitro splicing assay was utilized and transcripts derived from CES2 gene fragments of the wild type and IVS8-2A>G were compared. Sequence analysis of the cloned transcripts revealed that IVS8-2A>G yielded mostly aberrantly spliced transcripts, including a deleted exon or a 32-bp deletion proximal to the 5' end of exon 9, which resulted in truncated hCE-2 proteins. These results suggested that 100C>T (R34W), 424G>A (V142M), and IVS8-2A>G are functionally deficient SNPs.
Sigma-Aldrich
Carboxylesterase 2 human, recombinant, expressed in baculovirus infected BTI insect cells
Sigma-Aldrich
4-Methylumbelliferyl acetate, esterase substrate