Merck
  • Stem bromelain: an enzyme that naturally facilitates oriented immobilization.
Stem bromelain: an enzyme that naturally facilitates oriented immobilization.
Protein and peptide letters (2007-03-10)
Hafeeza Khatoon, Hina Younus, Mohammad Saleemuddin
ABSTRACT
The lone oligosaccharide chain of stem bromelain was oxidized with periodic acid to generate aldehyde groups and the resulting oxidized enzyme coupled to amino-Sepharose in order to obtain an immobilized preparation with uniformly oriented enzyme. The immobilized bromelain exhibited high proteolytic activity and remarkably enhanced thermal stability as compared to soluble bromelain and that coupled to CNBr activated Sepharose.
Sigma-Aldrich
Bromelain from pineapple stem, ≥4 units/mg protein, (chromatography purified)
Sigma-Aldrich
Bromelain from pineapple stem, ≥3 units/mg protein