Merck
Involvement of an ICE-like protease in Fas-mediated apoptosis.
Nature (1995-05-04)
M Enari, H Hug, S Nagata
ABSTRACT
Fas is a type-I membrane protein that transduces an apoptotic signal. Binding of Fas ligand or agonistic anti-Fas antibody to Fas kills the cells by apoptosis. Studies in the nematode Caenorhabditis elegans have suggested that proteases such as interleukin-1 beta-converting enzyme (ICE) or the product of the C. elegans cell-death gene ced-3 are involved in apoptotic signal transduction. The activity of ICE can be inhibited by the product of crmA, a cytokine-response modifier gene encoded by cowpox virus. We report here that expression of crmA inhibits cytotoxicity induced by anti-Fas antibody or tumour necrosis factor (TNF). We have found a specific ICE inhibitor tetrapeptide (acetyl-Tyr-Val-Ala-Asp-chloromethylketone) that also prevents apoptosis induced by anti-Fas antibody. These results suggest an involvement of an ICE-like protease in Fas-mediated apoptosis and TNF-induced cytotoxicity.