Merck
  • Extracellular K(+) and opening of voltage-gated potassium channels activate T cell integrin function: physical and functional association between Kv1.3 channels and beta1 integrins.
Extracellular K(+) and opening of voltage-gated potassium channels activate T cell integrin function: physical and functional association between Kv1.3 channels and beta1 integrins.
The Journal of experimental medicine (2000-04-05)
M Levite, L Cahalon, A Peretz, R Hershkoviz, A Sobko, A Ariel, R Desai, B Attali, O Lider
ABSTRACT
Elevated extracellular K(+) ([K(+)](o)), in the absence of "classical" immunological stimulatory signals, was found to itself be a sufficient stimulus to activate T cell beta1 integrin moieties, and to induce integrin-mediated adhesion and migration. Gating of T cell voltage-gated K(+) channels (Kv1.3) appears to be the crucial "decision-making" step, through which various physiological factors, including elevated [K(+)](o) levels, affect the T cell beta1 integrin function: opening of the channel leads to function, whereas its blockage prevents it. In support of this notion, we found that the proadhesive effects of the chemokine macrophage-inflammatory protein 1beta, the neuropeptide calcitonin gene-related peptide (CGRP), as well as elevated [K(+)](o) levels, are blocked by specific Kv1.3 channel blockers, and that the unique physiological ability of substance P to inhibit T cell adhesion correlates with Kv1.3 inhibition. Interestingly, the Kv1.3 channels and the beta1 integrins coimmunoprecipitate, suggesting that their physical association underlies their functional cooperation on the T cell surface. This study shows that T cells can be activated and driven to integrin function by a pathway that does not involve any of its specific receptors (i.e., by elevated [K(+)](o)). In addition, our results suggest that undesired T cell integrin function in a series of pathological conditions can be arrested by molecules that block the Kv1.3 channels.