Retinyl Acetate (Vitamin A Acetate)
Synonym(s):
Retinyl acetate, Retinol acetate, Vitamin A acetate, Retinyl acetate solution, Vitamin A acetate solution
Empirical Formula (Hill Notation):
C22H32O2
CAS Number:
Molecular Weight:
328.49
Products
Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing