Skip to Content
Merck
Aciclovir British Pharmacopoeia (BP) Reference Standard

Acyclovir

Aciclovir British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Acycloguanosine, 9-[(2-Hydroxyethoxy)methyl]guanine, Acyclovir
Empirical Formula (Hill Notation):
C8H11N5O3
CAS Number:
Molecular Weight:
225.20
EC Number:

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique