Potassium chloride solution
Synonym(s):
Potassium chloride solution saturated with AgCl, Potassium chloride solution
Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
Products
Brand
Formula Weight
pH Value
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Reaction Type
Reference Material Type
Special Grade
Technique
Showing 1-19 of 19
Compare
Product Number
Description
Pricing