Skip to Content
NL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Formic acid
Linear Formula:
HCOOH
CAS Number:
64-18-6
Molecular Weight:
46.03
Products
Product Category
acids (1)
organic acids (1)
solvents (1)
Brand
Sigma-Aldrich (1)
Supelco (3)
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade
ACS reagent (1)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
399388
Formic acid,
ACS reagent, ≥88%
Expand
View Pricing
1.59013
Formic acid,
hypergrade for LC-MS LiChrosolv
®
Expand
View Pricing
06473
Formic acid,
BioUltra, 1.0 M in H
2
O
Expand
View Pricing
1.00253
Formic acid,
for analysis EMSURE
®
ACS
Expand
View Pricing
1.10854
Formic acid,
for determination of viscosity acc. to DIN EN ISO 307
Expand
View Pricing