Skip to Content
Merck
Boron nitride Boron nitride. CAS 10043-11-5, pH 5 - 8 (100 g/l, H₂O, 20 °C) (slurry).

Hexagonal boron nitride ink

Boron nitride Boron nitride. CAS 10043-11-5, pH 5 - 8 (100 g/l, H₂O, 20 °C) (slurry).
Synonym(s):
Boron nitride
Empirical Formula (Hill Notation):
BN
CAS Number:
Molecular Weight:
24.82

Products

Brand
Sustainability
Boiling Point (°C)
Color
Feature
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Particle Size (µm)