Skip to Content
NL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Milliflex Cassette
Products
Product Category
media (7)
Brand
Millipore (7)
Color
Physical Form
Quality Segment
Application
Microorganism Suitability
Package Size
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
MXSMEND48
Milliflex Cassette,
Prefilled with m-Endo LES Agar designed for the recovery of coliform organisms
Expand
View Pricing
MXSMCSP48
Milliflex Cassette,
Prefilled with Sabouraud Dextrose Agar with Chloramphenicol designed for the recovery of a broad range of yeasts and molds
Expand
View Pricing
MXSMCET48
Milliflex Cassette,
Prefilled with Cetrimide Agar designed for the isolation and identification of
Pseudomonas aeruginosa
found in various types of water, especially high purity water
Expand
View Pricing
MXSMCMC24
Milliflex Cassette,
Prefilled with MacConkey Agar designed to detect Lactose fermenting organisms
Expand
View Pricing
MXSMKFS48
Milliflex Cassette,
Prefilled with KF
Streptococcus
Agar designed for the recovery of fecal
Streptococci
(enterococci)
Expand
View Pricing
MXSMPCA48
Milliflex Cassette,
Prefilled with Plate Count Agar designed for the recovery of bacteria (aerobic microorganisms)
Expand
View Pricing
MXSMTLP48
Milliflex Cassette,
Prefilled with Tryptic Soy Agar with Polysorbate 80 and Lecithin designed for the recovery of heterotrophic microorganisms
Expand
View Pricing