
Calcium Folinate Hydrate amorphous / Leucovorin Calcium amorphous

Synonym(s):
Folinic acid calcium salt, Calcium folinate
Empirical Formula (Hill Notation):
C20H21CaN7O7
CAS Number:
Molecular Weight:
511.50
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Agency
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing