Silicon dioxide
Synonym(s):
Cristobalite, Quartz, Sand, Silica
Linear Formula:
SiO2
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Purity
Length (mm)
Quality Segment
Application
Special Grade
Showing 1-6 of 6
Compare
Product Number
Description
Pricing