Skip to Content
Merck
Quercetin 3-β-D-glucoside ≥90% (HPLC)

Quercetin 3-β-D-glucoside

Quercetin 3-β-D-glucoside ≥90% (HPLC)
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 3-β-glucoside, Isoquercitrin
Empirical Formula (Hill Notation):
C21H20O12
CAS Number:
Molecular Weight:
464.38

Products

Product Category
Brand
Formula Weight
Gene Alias
Purity
Research Area
Quality Segment
Application
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing