Skip to Content
Merck
Ibuprofen meets USP testing specifications

Ibuprofen

Ibuprofen meets USP testing specifications
Synonym(s):
α-Methyl-4-(isobutyl)phenylacetic acid, (±)-2-(4-Isobutylphenyl)propanoic acid
Empirical Formula (Hill Notation):
C13H18O2
CAS Number:
Molecular Weight:
206.28
EC Number:

Products

Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Gene Alias
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing