Skip to Content
Merck
Poly(ethylene glycol) analytical standard, for GPC, 600

Poly(ethylene glycol)

Poly(ethylene glycol) analytical standard, for GPC, 600
Synonym(s):
PEG, Poly(ethylene glycol), PEO
Linear Formula:
H(OCH2CH2)nOH
CAS Number:

Products

Product Category
Brand
Sustainability
Feature
Manufacturer
Melting Point (°C)
pH Value
Physical Form
Product Line
Agency
Format
Quality Segment
Functional Group
Application
Polymer Composition
Polymer Type
API Family
Reference Material Type
Special Grade
Technique