Polymer Synthesis Techniques
Polymers are a class of materials comprising small repeating chemical units. The building block or the repeating unit in the polymer is called a monomer. Polymerization, or polymer synthesis, is a chemical reaction in which monomers are joined together by covalent bonding to form polymer structures. The length of the polymer chain is characterized by the number of repeating units in the chain, which is referred to as degree of polymerization (DP). The molecular weight of the polymer is the product of the molecular weight of the repeating unit and the DP. The basic properties inherent to polymers mainly depend on the molecular weight, structure (linear or branched), and DP.
Featured Categories
We provide reliable polymerization tools to initiate and manipulate a chemical reaction of monomers to form polymer chains or networks with specific properties for diverse applications.
Discover our selection of polyethylene glycol (PEGs) and PEG derivatives in a wide range of molecular weights for all your PEGylation needs and applications.
We offer an extensive range of natural and synthetic biomedical polymers with advanced properties suitable for all your biomedical applications.
Explore our portfolio of versatile hydrophobic polymers to support your adhesion, coating, fiber, film, engineered plastic and biomedical applications.