- Differential transcriptome of tolerogenic versus inflammatory dendritic cells points to modulated T1D genetic risk and enriched immune regulation.
Differential transcriptome of tolerogenic versus inflammatory dendritic cells points to modulated T1D genetic risk and enriched immune regulation.
Genes and immunity (2017-08-11)
T Nikolic, N J C Woittiez, A van der Slik, S Laban, A Joosten, C Gysemans, C Mathieu, J J Zwaginga, B Koeleman, B O Roep
PMID28794505
ABSTRACT
Tolerogenic dendritic cells (tolDCs) are assessed as immunomodulatory adjuvants to regulate autoimmunity. The underlying gene expression endorsing their regulatory features remains ill-defined. Using deep mRNA sequencing, we compared transcriptomes of 1,25-dihydroxyvitaminD