Merck
  • Influence of iodine in excess on seminiferous tubular structure and epididymal sperm character in male rats.
Influence of iodine in excess on seminiferous tubular structure and epididymal sperm character in male rats.
Environmental toxicology (2017-02-17)
Amar K Chandra, Arijit Chakraborty
ABSTRACT
Excess iodine induced public health problems are now emerging in many iodine sufficient regions for indiscriminate intake of iodine through various iodized products. It has been reported that excess iodine can disrupt overall male reproductive physiology by generating oxidative stress in the testis. However, information on the possible effect of iodine in excess on spermatozoa found less. In the present investigation flow cytometric techniques and scanning electron microscopy (SEM) have been used to study the spermatozoal functional as well as structural status under the influence of excess iodine; generation of ROS in the spermatozoa as evident by DCFDA, altered acrosomal integrity as observed by fluorescence lectin staining method and depolarized mitochondrial membrane potential (ΔΨ