- TRPV4 calcium entry channel: a paradigm for gating diversity.
TRPV4 calcium entry channel: a paradigm for gating diversity.
American journal of physiology. Cell physiology (2004-01-07)
Bernd Nilius, Joris Vriens, Jean Prenen, Guy Droogmans, Thomas Voets
PMID14707014
ABSTRACT
The vanilloid receptor-1 (VR1, now TRPV1) was the founding member of a subgroup of cation channels within the TRP family. The TRPV subgroup contains six mammalian members, which all function as Ca2+ entry channels gated by a variety of physical and chemical stimuli. TRPV4, which displays 45% sequence identity with TRPV1, is characterized by a surprising gating promiscuity: it is activated by hypotonic cell swelling, heat, synthetic 4alpha-phorbols, and several endogenous substances including arachidonic acid (AA), the endocannabinoids anandamide and 2-AG, and cytochrome P-450 metabolites of AA, such as epoxyeicosatrienoic acids. This review summarizes data on TRPV4 as a paradigm of gating diversity in this subfamily of Ca2+ entry channels.