Merck
  • Are patients with rheumatoid arthritis still at an increased risk of tuberculosis and what is the role of biological treatments?
Are patients with rheumatoid arthritis still at an increased risk of tuberculosis and what is the role of biological treatments?
Annals of the rheumatic diseases (2014-03-13)
Elizabeth V Arkema, Jerker Jonsson, Eva Baecklund, Judith Bruchfeld, Nils Feltelius, Johan Askling
ABSTRACT
To estimate the risk of tuberculosis (TB) in patients with rheumatoid arthritis (RA) both with and without exposure to biological therapy and to directly compare the risks between therapies. Data from the Swedish National Population Registers, Tuberculosis Register and the Swedish Biologics Register were used to conduct a prospective population-based national cohort study (2002-2011). We estimated the rate of incident TB in the general population and in a cohort of biological-naïve and biological-exposed patients diagnosed with RA. Cox models were used to estimate HRs with particular attention to risks by calendar and follow-up time and individual biologics. Compared to the general population, RA patients not exposed to biologicals had a fourfold increased risk of TB (HR 4.2; 95% CI 2.7 to 6.7), which did not decline over calendar time. In contrast, the risk of TB in the biological-exposed RA population decreased since 2002 compared with biological-naïve; from HR=7.9 (95% CI 3.3 to 18.9) in 2002-2006 to HR=2.4 (95% CI 0.9 to 6.1) in 2007-2011. The HRs for most recent exposure to adalimumab and infliximab compared with etanercept were 3.1 (95% CI 0.8 to 12.5) and 2.7 (95% CI 0.7 to 10.9), respectively, and the HR for etanercept compared with biological-naïve RA was 1.7 (95% CI 0.6 to 4.6). In the past decade, the risk of TB has decreased among biological-exposed RA patients but remains higher than in biological-naïve RA patients. Most cases of TB in RA occur in biological-naïve RA patients, underscoring the elevated risk also in these patients.
Product Description
Supelco
Isoniazid, Vetec, reagent grade, 98%