Merck
  • Inducible brown adipocytes in subcutaneous inguinal white fat: the role of continuous sympathetic stimulation.
Inducible brown adipocytes in subcutaneous inguinal white fat: the role of continuous sympathetic stimulation.
American journal of physiology. Endocrinology and metabolism (2014-09-04)
G Andres Contreras, Yun-Hee Lee, Emilio P Mottillo, James G Granneman
ABSTRACT
Brown adipocytes (BA) generate heat in response to sympathetic activation and are the main site of nonshivering thermogenesis in mammals. Although most BA are located in classic brown adipose tissue depots, BA are also abundant in the inguinal white adipose tissue (iWAT) before weaning. The number of BA is correlated with the density of sympathetic innervation in iWAT; however, the role of continuous sympathetic tone in the establishment and maintenance of BA in WAT has not been investigated. BA marker expression in iWAT was abundant in weaning mice but was greatly reduced by 8 wk of age. Nonetheless, BA phenotype could be rapidly reinstated by acute β₃-adrenergic stimulation with CL-316,243 (CL). Genetic tagging of adipocytes with adiponectin-CreER(T2) demonstrated that CL reinstates uncoupling protein 1 (UCP1) expression in adipocytes that were present before weaning. Chronic surgical denervation dramatically reduced the ability of CL to induce the expression of UCP1 and other BA markers in the tissue as a whole, and this loss of responsiveness was prevented by concurrent treatment with CL. These results indicate that ongoing sympathetic activity is critical to preserve the ability of iWAT fat cells to express a BA phenotype upon adrenergic stimulation.