- Macrophage-specific expression of mannose-binding lectin controls atherosclerosis in low-density lipoprotein receptor-deficient mice.
Macrophage-specific expression of mannose-binding lectin controls atherosclerosis in low-density lipoprotein receptor-deficient mice.
With consideration of the central role of the innate immune system in atherogenesis and mannose-binding lectin (MBL) as an innate regulator of immunity, the role of MBL in experimental and human atherosclerosis was assessed. With the use of immunohistochemistry and polymerase chain reaction, deposition and gene expression of MBL-A and -C were assessed in murine atherosclerosis from mice deficient for the low-density lipoprotein receptor (LDLR(-/-)) after 10 or 18 weeks of high-fat feeding. MBL was present and was produced in 10-week-old lesions, whereas deposition and gene expression were minimal after 18 weeks of high-fat feeding and absent in healthy vasculature. Interestingly, deposition of MBL-A and -C differed: MBL-A predominantly localized in upper medial layers, whereas MBL-C was found in and around intimal macrophages. To further study the role of local MBL production by monocytic cells in atherosclerosis, LDLR(-/-) mice with MBL-A and -C(-/-) monocytic cells were construed by bone marrow transplantation. Mice carrying MBL-A and -C double deficient macrophages had increased (30%) atherosclerotic lesions compared with wild-type controls (P=0.015) after 10 weeks of high-fat diet. Subsequently, analysis of MBL deposition and gene expression in advanced human atherosclerotic lesions revealed the presence of MBL protein in ruptured but not stable atherosclerotic lesions. Putatively in agreement with murine data, no MBL gene expression could be detected in advanced human atherosclerotic lesions. These results are the first to show that MBL is abundantly present and locally produced during early atherogenesis. Local MBL expression, by myeloid cells, is shown to critically control development of atherosclerotic lesions.