- Perineal muco-cutaneous herpes zoster treated with brivudin.
Perineal muco-cutaneous herpes zoster treated with brivudin.
The Journal of dermatological treatment (2008-07-09)
Osvaldo Correia, Marta A Pereira, Teresa Pereira, António Santos
PMID18608718
ABSTRACT
Herpes zoster results from the reactivation of latent varicella-zoster virus (VZV) from the dorsal root ganglion of sensory nerves. It is rarely described in the pediatric population. We report the case of an 8-year-old immunocompetent boy with a painful lumbosacral herpes zoster that was treated with brivudin and achieved rapid and sustained improvement in the absence of muco-cutaneous or pharmacological side effects.