Merck
In vitro generation of lymphocytes from embryonic stem cells.
Methods in molecular biology (Clifton, N.J.) (2004-09-14)
Renée F de Pooter, Sarah K Cho, Juan Carlos Zúñiga-Pflücker
ABSTRACT
Lymphocytes arise during ontogeny via a series of increasingly restricted intermediates. Initially, the mesoderm gives rise to hemangioblasts, which can differentiate into endothelial precursors, or hematopoietic stem cells (HSCs). HSCs can either self-renew or differentiate into lineage-restricted progenitors and, ultimately, to mature effector cells. This complex process is only beginning to be understood, and the ability to generate lymphocytes from embryonic stem (ES) cells in vitro will facilitate further study by providing a model system in which the effects of genetic and environmental manipulations of ES-cell-derived progenitors can be examined. In this protocol, we describe procedures for generating either B- and NK- or T-lymphocytes from mouse ES cells in vitro.