Merck
  • SARS-CoV-2-triggered mast cell rapid degranulation induces alveolar epithelial inflammation and lung injury.
SARS-CoV-2-triggered mast cell rapid degranulation induces alveolar epithelial inflammation and lung injury.
Signal transduction and targeted therapy (2021-12-19)
Meng-Li Wu, Feng-Liang Liu, Jing Sun, Xin Li, Xiao-Yan He, Hong-Yi Zheng, Yan-Heng Zhou, Qihong Yan, Ling Chen, Guo-Ying Yu, Junbiao Chang, Xia Jin, Jincun Zhao, Xin-Wen Chen, Yong-Tang Zheng, Jian-Hua Wang
ABSTRACT
SARS-CoV-2 infection-induced hyper-inflammation links to the acute lung injury and COVID-19 severity. Identifying the primary mediators that initiate the uncontrolled hypercytokinemia is essential for treatments. Mast cells (MCs) are strategically located at the mucosa and beneficially or detrimentally regulate immune inflammations. In this study, we showed that SARS-CoV-2-triggered MC degranulation initiated alveolar epithelial inflammation and lung injury. SARS-CoV-2 challenge induced MC degranulation in ACE-2 humanized mice and rhesus macaques, and a rapid MC degranulation could be recapitulated with Spike-RBD binding to ACE2 in cells; MC degranulation altered various signaling pathways in alveolar epithelial cells, particularly, the induction of pro-inflammatory factors and consequential disruption of tight junctions. Importantly, the administration of clinical MC stabilizers for blocking degranulation dampened SARS-CoV-2-induced production of pro-inflammatory factors and prevented lung injury. These findings uncover a novel mechanism for SARS-CoV-2 initiating lung inflammation, and suggest an off-label use of MC stabilizer as immunomodulators for COVID-19 treatments.