Merck
  • Flavine adenine dinucleotide inhibits pathological cardiac hypertrophy and fibrosis through activating short chain acyl-CoA dehydrogenase.
Flavine adenine dinucleotide inhibits pathological cardiac hypertrophy and fibrosis through activating short chain acyl-CoA dehydrogenase.
Biochemical pharmacology (2020-06-17)
Zhichao Ma, Xue Qin, Xiaoyi Zhong, Yingqing Liao, Yongshao Su, Xi Liu, Peiqing Liu, Jing Lu, Sigui Zhou
ABSTRACT
Short-chain acyl-CoA dehydrogenase (SCAD), the rate-limiting enzyme for fatty acid β-oxidation, has a negative regulatory effect on pathological cardiac hypertrophy and fibrosis. Furthermore, flavin adenine dinucleotide (FAD) can enhance the expression and enzyme activity of SCAD. However, whether FAD can inhibit pathological cardiac hypertrophy and fibrosis remains unclear. Therefore, we observed the effect of FAD on pathological cardiac hypertrophy and fibrosis. FAD significantly inhibited PE-induced cardiomyocyte hypertrophy and AngII-induced cardiac fibroblast proliferation. In addition, FAD ameliorated pathological cardiac hypertrophy and fibrosis in SHR. FAD significantly increased the expression and enzyme activity of SCAD. Meanwhile, ATP content was increased, the content of free fatty acids and reactive oxygen species were decreased by FAD in vivo and in vitro. In addition, molecular dynamics simulations were also used to provide insights into the structural stability and dynamic behavior of SCAD. The results demonstrated that FAD may play an important structural role on the SCAD dimer stability and maintenance of substrate catalytic pocket to increase the expression and enzyme activity of SCAD. In conclusion, FAD can inhibit pathological cardiac hypertrophy and fibrosis through activating SCAD, which may be a novel effective treatment for pathological cardiac hypertrophy and fibrosis, thus prevent them from developing into heart failure.
Sigma-Aldrich
(+)-Biotin 4-nitrophenyl ester, 98%