Merck
  • Study of Biointegration and Elastic-Strength Properties of a New Xenopericardium-Based Biomaterial for Reconstructive Cardiovascular Surgery.
Study of Biointegration and Elastic-Strength Properties of a New Xenopericardium-Based Biomaterial for Reconstructive Cardiovascular Surgery.
Bulletin of experimental biology and medicine (2019-09-09)
I S Fadeeva, M N Sorkomov, A I Zvyagina, D V Britikov, A S Sachkov, Ya V Evstratova, R S Fadeev, R M Muratov, V S Akatov
ABSTRACT
We analyzed biocompatibility, elastic-strength properties, and biointegration potential of a new biomaterial made of xenopericardium for reconstructive cardiovascular surgery. The biomaterial manufactured by the proposed technology demonstrated high biocompatibility and biointegration potential and its elastic-strength properties 2-4-fold surpassed that of native pericardium. The obtained results attested to good prospects of using the proposed technology for preparing biomaterials for reconstructive cardiovascular surgery.
Sigma-Aldrich
Sodium deoxycholate monohydrate, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Sodium deoxycholate monohydrate, BioXtra, ≥99.0% (titration)