- Monoamine oxidase activity in noradrenaline neurons of the locus coeruleus of the rat. A double-labeling histochemical study.
Monoamine oxidase activity in noradrenaline neurons of the locus coeruleus of the rat. A double-labeling histochemical study.
The aim of the present study is to examine whether noradrenergic neurons of the locus coeruleus (LC) of the rat contain monoamine oxidase (MAO) activity. Sections were processed initially for MAO enzyme histochemistry using tyramine as a substrate, followed by fluorescence immunohistochemistry for tyrosine hydroxylase (TH). In the LC, virtually all TH-immunoreactive neurons (i.e., noradrenergic neurons) were also positive for MAO. No MAO activity was found in any TH-negative neurons. Neurons in the LC have previously been shown to form dopamine during noradrenaline biosynthesis and to produce serotonin from exogenously administered l-5-hydroxytryptophan. Moreover, dopamine- and serotonin-degrading MAO activity has also been found in LC neurons. Therefore, our results indicate that MAO activity is localized within noradrenergic neurons in the LC and is likely involved in the degradation of dopamine that is endogenously synthesized, and also in the elimination of serotonin that is produced from exogenous precursors.