Merck
  • The calcitonin receptor stimulates Shc tyrosine phosphorylation and Erk1/2 activation. Involvement of Gi, protein kinase C, and calcium.
The calcitonin receptor stimulates Shc tyrosine phosphorylation and Erk1/2 activation. Involvement of Gi, protein kinase C, and calcium.
The Journal of biological chemistry (1998-07-25)
Y Chen, J F Shyu, A Santhanagopal, D Inoue, J P David, S J Dixon, W C Horne, R Baron
ABSTRACT
While it is well established that adenylyl cyclase and phospholipase C-beta are two proximal signal effectors for the calcitonin receptor, the more distal signaling pathways are less well characterized. G protein-coupled receptors can activate Erk1/2 by Gs-, Gi-, or Gq-dependent signaling pathways, depending on the specific receptor and cell type examined. Since the calcitonin receptor can couple to all three of these G proteins, the ability of calcitonin to activate Erk1/2 was investigated. Calcitonin induced time- and concentration-dependent increases in Shc tyrosine phosphorylation, Shc-Grb2 association and Erk1/2 phosphorylation and activation in a HEK 293 cell line that stably expresses the rabbit calcitonin receptor C1a isoform. Pertussis toxin, which inactivates Gi, and calphostin C, a protein kinase C inhibitor, each partially inhibited calcitonin-induced Shc tyrosine phosphorylation, Shc-Grb2 association, and Erk1/2 phosphorylation. In contrast, neither forskolin nor H89, a protein kinase A inhibitor, had a significant effect on basal or calcitonin-stimulated Erk1/2 phosphorylation. Our results suggest that the calcitonin receptor induces Shc phosphorylation and Erk1/2 activation in HEK293 cells by parallel Gi- and PKC-dependent mechanisms. The calcitonin-induced elevation of cytosolic free Ca2+ was required for Erk1/2 phosphorylation, since preventing any change in cytosolic free Ca2+ by chelating both cytosolic and extracellular Ca2+ abolished the response. However, the change in Ca2+ that is induced by calcitonin is not sufficient to account for the calcitonin-induced Erk1/2 phosphorylation, since treatment with 100 nM ionomycin or 10 microM thapsigargin, each of which induced elevations of Ca2+ comparable to those induced by calcitonin, induced significantly less Erk1/2 phosphorylation than that induced by calcitonin. Erk1/2 may have important roles as downstream effectors mediating cellular responses to calcitonin stimulation.