- HSF1, in association with MORC2, downregulates ArgBP2 via the PRC2 family in gastric cancer cells.
HSF1, in association with MORC2, downregulates ArgBP2 via the PRC2 family in gastric cancer cells.
Arg Kinase-binding protein 2 (ArgBP2) is considered to be a scaffold protein that coordinates multiple signaling pathways converging on cell adhesion and actin cytoskeletal organization. It also plays an important role in blocking cancer metastasis as a potential tumor suppressor. However, its regulation mechanisms in tumor migration, especially in gastric cancer, are not fully understood. Here, we identified an ArgBP2 enhancer and showed that heat shock factor 1 (HSF1) directly interacted with microrchidia CW-type zinc finger 2 (MORC2) and bound to the enhancer of ArgBP2. HSF1 was found to promote proliferation, migration and invasion of gastric cancer cells. HSF1 or/and MORC2 increased recruitment of the polycomb repressive complex 2 (PRC2), particularly enhancer of zeste homolog 2 (EZH2), to the ArgBP2 enhancer and catalyzed tri-methylation of lysine 27 on histone H3 (H3K27me3), leading to transcriptional repression of ArgBP2. In addition, HSF1 and MORC2-induced migration and invasion in gastric cancer cells was dependent on ArgBP2 or EZH2. Clinical data exhibited a negative correlation of ArgBP2 with MORC2, HSF1, and EZH2. Our results thus contribute to the knowledge of the regulatory mechanism of HSF1 in down-regulating ArgBP2, providing new insight into the HSF1&MORC2-PRC2-ArgBP2 signaling pathway and a better understanding of their functions in gastric cancer cells.